Ethyl pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-pyrrole-2-carboxylate

A262281-100G A262281-25G A262281-500G A262281-5G A262281-10G
76771-898EA 91 USD
76771-898 76771-900 76771-902 76771-904 77626-370
Ethyl pyrrole-2-carboxylate 97%
Ethyl pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD00817049
CAS Number: 2199-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR