6-Iodo-3,4-dihydronaphthalen-1(2H)-one 98%

Supplier: Ambeed
Warning

A261860-1G A261860-250MG A261860-5G
76771-892EA 33 USD
76771-892 76771-894 77568-512
6-Iodo-3,4-dihydronaphthalen-1(2H)-one 98%
6-Iodo-3,4-dihydronaphthalen-1(2H)-one

Precaución: For research use only.

Formula: C₁₀H₉IO
MDL Number: MFCD09751536
CAS Number: 340825-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR