5-Methyl-8-quinolinol 98%

Supplier: Ambeed
Warning

Synonyms: Tiliquinol, 5-Methylquinolin-8-ol

A260708-1G A260708-250MG A260708-5G A260708-100MG A260708-25G
76771-858EA 45.62 USD
76771-858 76771-860 77571-610 77626-358 77626-360
5-Methyl-8-quinolinol 98%
5-Methyl-8-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00864652
CAS Number: 5541-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR