Methyl-2-amino-4-methylthiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-4-methylthiazole-5-carboxylate

A260235-100G A260235-1G A260235-25G A260235-5G
76771-850EA 712.93 USD
76771-850 76771-852 76771-854 76771-856
Methyl-2-amino-4-methylthiazole-5-carboxylate 97%
Methyl 2-amino-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.2049 g/mol
MDL Number: MFCD01993690
CAS Number: 3829-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR