5-Chloro-2,2-difluoro-1,3-benzodioxole 98%

Supplier: Ambeed
Warning

A259629-1G A259629-250MG A259629-5G
76771-840EA 51.99 USD
76771-840 76771-842 76771-844
5-Chloro-2,2-difluoro-1,3-benzodioxole 98%
5-Chloro-2,2-difluoro-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.54 g/mol
MDL Number: MFCD04972706
CAS Number: 72769-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR