2'-Amino-4',5'-dimethoxyacetophenone 96%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Amino-4,5-dimethoxyphenyl)ethanone

A259017-1G A259017-250MG A259017-25G A259017-5G A259017-100G
76771-832EA 8.49 USD
76771-832 76771-834 76771-836 76771-838 77626-348
2'-Amino-4',5'-dimethoxyacetophenone 96%
2'-Amino-4',5'-dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD00016646
CAS Number: 4101-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR