5-Chloro-2-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

A258441-100G A258441-25G A258441-5G
76771-820EA 76.39 USD
76771-820 76771-822 76771-824
5-Chloro-2-(trifluoromethoxy)aniline 98%
5-Chloro-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD06410919
CAS Number: 326-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR