N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Chloro-4-fluoroanilino)-7-fluoro-6-nitroquinazoline

A257382-100G A257382-1G A257382-25G A257382-5G A257382-10G
76771-802EA 219.61 USD
76771-802 76771-804 76771-806 76771-808 77598-364
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine 98%
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine

Precaución: for Research Use Only

Formula: C₁₄H₇ClF₂N₄O₂
MW: 336.68 g/mol
MDL Number: MFCD09998825
CAS Number: 162012-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR