2-Bromo-3-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A255963-10G A255963-1G A255963-25G A255963-5G
76771-778EA 70 USD
76771-778 76771-780 76771-782 76771-784
2-Bromo-3-fluorobenzonitrile 98%
2-Bromo-3-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrFN
MDL Number: MFCD07368773
CAS Number: 425379-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR