4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: N4-Boc-piperazine-2-carboxylic acid

A254530-1G A254530-250MG A254530-10G A254530-25G
76771-766EA 7.43 USD
76771-766 76771-768 77626-336 77626-338
4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%
4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.26 g/mol
MDL Number: MFCD01076218
CAS Number: 128019-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR