Methyl-5-nitro-1H-pyrazole-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-nitro-1H-pyrazole-3-carboxylate
A254029-100MG
A254029-10G
A254029-1G
A254029-250MG
A254029-25G
A254029-5G
76771-750EA
22.28
USD
76771-750
76771-752
76771-754
76771-756
76771-758
76771-760
Methyl-5-nitro-1H-pyrazole-3-carboxylate 98%
Methyl-5-nitro-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₅H₅N₃O₄ MW: 171.1100 g/mol |
MDL Number:
MFCD08236713 CAS Number: 181585-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...