2-Bromo-4-nitro(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-nitro-1-(trifluoromethoxy)benzene

A253465-100G A253465-10G A253465-1G A253465-25G A253465-500G A253465-5G
76771-720EA 95 USD
76771-720 76771-722 76771-724 76771-726 76771-728 76771-730
2-Bromo-4-nitro(trifluoromethoxy)benzene 98%
2-Bromo-4-nitro(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₃
MDL Number: MFCD04973758
CAS Number: 200958-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR