6-Chloro-2-methylquinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

A253070-1G A253070-250MG
76771-710EA 22 USD
76771-710 76771-712
6-Chloro-2-methylquinazolin-4(3H)-one 97%
6-Chloro-2-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD00195330
CAS Number: 7142-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR