2,5-Dichlorobenzonitrile 98%

Supplier: Ambeed
Danger

A252915-25G A252915-5G
76771-706EA 45.62 USD
76771-706 76771-708
2,5-Dichlorobenzonitrile 98%
2,5-Dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
MDL Number: MFCD00019741
CAS Number: 21663-61-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR