5-Chloro-2-mercaptobenzothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chlorobenzo[d]thiazole-2(3H)-thione

A252585-100G A252585-25G A252585-5G
76771-696EA 476.35 USD
76771-696 76771-698 76771-700
5-Chloro-2-mercaptobenzothiazole 95%
5-Chloro-2-mercaptobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₄ClNS₂
MW: 201.7 g/mol
MDL Number: MFCD00005783
CAS Number: 5331-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR