Ethyl-N-(2-cyanoacetyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Cyanoacetylurethane

A252429-100G A252429-10G A252429-1G A252429-25G A252429-5G
76771-674EA 143 USD
76771-674 76771-676 76771-678 76771-680 76771-682
Ethyl-N-(2-cyanoacetyl)carbamate 95%
Ethyl-N-(2-cyanoacetyl)carbamate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
MDL Number: MFCD00001937
CAS Number: 6629-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR