4-Bromo-4'-heptylbiphenyl 97%

Supplier: Ambeed
Warning

A252263-1G A252263-25G A252263-5G
76771-668EA 14 USD
76771-668 76771-670 76771-672
4-Bromo-4'-heptylbiphenyl 97%
4-Bromo-4'-heptylbiphenyl

Precaución: for Research Use Only

Formula: C₁₉H₂₃Br
MDL Number: MFCD00060107
CAS Number: 58573-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR