5,6-Dichloro-2-mercaptobenzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-1H-benzimidazole-2-thiol

A251732-250MG A251732-5G A251732-1G
76771-664EA 19.1 USD
76771-664 76771-666 77593-584
5,6-Dichloro-2-mercaptobenzimidazole 97%
5,6-Dichloro-2-mercaptobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂S
MW: 219.09 g/mol
MDL Number: MFCD00052398
CAS Number: 19462-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR