2-Fluoro-3-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene

A251565-100G A251565-10G A251565-25G A251565-500G A251565-5G
76771-654EA 63 USD
76771-654 76771-656 76771-658 76771-660 76771-662
2-Fluoro-3-nitrotoluene 98%
2-Fluoro-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD03412242
CAS Number: 437-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR