4-Methyl-1H-imidazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A250221-1G A250221-250MG A250221-25G
76771-634EA 19 USD
76771-634 76771-636 77877-839
4-Methyl-1H-imidazole-5-carboxylic acid 97%
4-Methyl-1H-imidazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MDL Number: MFCD00233368
CAS Number: 1457-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR