4,6-Difluoroindole 96%

Supplier: Ambeed
Warning

A249528-100G A249528-1G A249528-250MG A249528-25G A249528-5G
76771-610EA 2085.73 USD
76771-610 76771-612 76771-614 76771-616 76771-618
4,6-Difluoroindole 96%
4,6-Difluoroindole

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD01075213
CAS Number: 199526-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR