4-Fluoro-3-iodotoluene 97%

Supplier: Ambeed
Warning

A248983-100G A248983-10G A248983-1G A248983-25G
76771-596EA 75 USD
76771-596 76771-598 76771-600 76771-602
4-Fluoro-3-iodotoluene 97%
4-Fluoro-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆FI
MDL Number: MFCD00079759
CAS Number: 452-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR