2,4-Dimethoxycinnamic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4-Dimethoxyphenyl)acrylic acid

A248265-10G A248265-1G A248265-5G
76771-584EA 106.09 USD
76771-584 76771-586 76771-588
2,4-Dimethoxycinnamic acid 95%
2,4-Dimethoxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00016837
CAS Number: 16909-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR