5-Fluoro-1H-benzimidazole 98%

Supplier: Ambeed
Warning

A247883-1G A247883-250MG A247883-5G
76771-574EA 14 USD
76771-574 76771-576 76771-578
5-Fluoro-1H-benzimidazole 98%
5-Fluoro-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MDL Number: MFCD00792438
CAS Number: 1977-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR