5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%

Supplier: Ambeed
Warning

A246782-100G A246782-10G A246782-1G A246782-25G A246782-5G
76771-554EA 219 USD
76771-554 76771-556 76771-558 76771-560 76771-562 76771-564
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Precaución: for Research Use Only

Formula: C₇H₅N₃O₃
MDL Number: MFCD00220274
CAS Number: 93-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR