3',5'-Dichloro-2'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Dichloro-2-fluorophenyl)ethanone

A246543-1G A246543-5G
76771-550EA 22.28 USD
76771-550 76771-552
3',5'-Dichloro-2'-fluoroacetophenone 98%
3',5'-Dichloro-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO
MW: 207.029 g/mol
MDL Number: MFCD03427276
CAS Number: 480438-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR