(3,4-Difluorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A246046-10G A246046-1G A246046-25G A246046-5G
76771-538EA 29 USD
76771-538 76771-540 76771-542 77877-822
(3,4-Difluorophenyl)acetonitrile 98%
(3,4-Difluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MDL Number: MFCD00009974
CAS Number: 658-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR