(S)-3-(Boc-amino)piperidine 97%

Supplier: Ambeed
Warning

A245850-1G A245850-250MG A245850-500G A245850-5G A245850-25G A245850-100G
76771-530EA 6 USD
76771-530 76771-532 76771-534 76771-536 77550-580 77598-362
(S)-3-(Boc-amino)piperidine 97%
(S)-3-(Boc-amino)piperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD03093383
CAS Number: 216854-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR