5-Nitro-1H-indazole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A245031-1G A245031-250MG
76771-524EA 152.77 USD
76771-524 76771-526
5-Nitro-1H-indazole-3-carbaldehyde 98%
5-Nitro-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅N₃O₃
MW: 191.15 g/mol
MDL Number: MFCD07781436
CAS Number: 677702-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR