2-(5-Chloro-2-phenoxyphenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
A244634-1G
A244634-25G
A244634-5G
76771-508EA
11
USD
76771-508
76771-510
76771-512
2-(5-Chloro-2-phenoxyphenyl)acetic acid 97%
2-(5-Chloro-2-phenoxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₁ClO₃ |
MDL Number:
MFCD07368830 CAS Number: 70958-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...