5-Bromo-1H-indazol-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1,2-dihydro-3H-indazol-3-one

A244600-10G A244600-1G A244600-250MG A244600-5G
76771-500EA 332.07 USD
76771-500 76771-502 76771-504 76771-506
5-Bromo-1H-indazol-3-ol 98%
5-Bromo-1H-indazol-3-ol

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.0300 g/mol
MDL Number: MFCD07781638
CAS Number: 7364-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR