Dimethyl-5-chloroisophthalate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloroisophthalic acid dimethyl ester, Dimethyl 5-chloroisophthalate

A243993-10G A243993-1G A243993-25G A243993-5G
76771-472EA 53 USD
76771-472 76771-474 76771-476 76771-478
Dimethyl-5-chloroisophthalate 98%
Dimethyl-5-chloroisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₄
MDL Number: MFCD00079752
CAS Number: 20330-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR