5-(Chloromethyl)uracil 97%

Supplier: Ambeed
Danger

Synonyms: 5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

A243718-100MG A243718-1G A243718-250MG A243718-5G
76771-456EA 16 USD
76771-456 76771-458 76771-460 76771-462
5-(Chloromethyl)uracil 97%
5-(Chloromethyl)uracil

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD00218445
CAS Number: 3590-48-5
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR