5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde
A242105-1G
A242105-250MG
A242105-5G
A242105-25G
76771-446EA
44.56
USD
76771-446
76771-448
76771-450
77626-294
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 97%
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₉H₁₁ClN₂O MW: 220.65 g/mol |
MDL Number:
MFCD00100755 CAS Number: 947-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...