Thiamine hydrochloride 99+%

Supplier: Ambeed
Warning

A241911-1000G A241911-100G A241911-10G A241911-1G A241911-25G A241911-500G
76771-430EA 108 USD
76771-430 76771-432 76771-434 76771-436 77090-408 77568-350
Thiamine hydrochloride 99+%
Thiamine hydrochloride

Precaución: For research use only.

Formula: C₁₂H₁₈Cl₂N₄OS
MDL Number: MFCD00012780
CAS Number: 67-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR