6-Chloro-5-iodonicotinic acid 96%
Supplier: Ambeed
|
Warning
|
A241071-100MG
A241071-1G
A241071-250MG
A241071-25G
A241071-5G
76771-410EA
13.8
USD
76771-410
76771-412
76771-414
76771-416
76771-418
6-Chloro-5-iodonicotinic acid 96%
6-Chloro-5-iodonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃ClINO₂ MW: 283.4510 g/mol |
MDL Number:
MFCD11100073 CAS Number: 59782-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...