Ethyl-3-cyclopropyl-3-oxopropanoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-cyclopropyl-3-oxopropanoate

A240404-100G A240404-10G A240404-25G A240404-500G A240404-5G A240404-1G
76771-392EA 45.62 USD
76771-392 76771-394 76771-396 76771-398 76771-400 77626-276
Ethyl-3-cyclopropyl-3-oxopropanoate 97%
Ethyl-3-cyclopropyl-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
MDL Number: MFCD00184570
CAS Number: 24922-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR