1-Propylpiperazine 97%

Supplier: Ambeed
Warning

A239891-100MG A239891-1G A239891-250MG A239891-5G
76771-378EA 18.04 USD
76771-378 76771-380 76771-382 76771-384
1-Propylpiperazine 97%
1-Propylpiperazine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 128.21 g/mol
MDL Number: MFCD00044591
CAS Number: 21867-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR