5-Chloro-3-cyanopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloronicotinonitrile

A237899-100G A237899-1G A237899-250MG
76771-334EA 1425.85 USD
76771-334 76771-336 76771-338
5-Chloro-3-cyanopyridine 98%
5-Chloro-3-cyanopyridine

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
MDL Number: MFCD03411334
CAS Number: 51269-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR