(1-Methyl-1H-pyrazol-5-yl)methanol 98%

Supplier: Ambeed
Warning

A237158-100MG A237158-1G A237158-250MG A237158-25G A237158-5G
76771-310EA 6 USD
76771-310 76771-312 76771-314 76771-316 76771-318
(1-Methyl-1H-pyrazol-5-yl)methanol 98%
(1-Methyl-1H-pyrazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₈N₂O
MDL Number: MFCD08556301
CAS Number: 84547-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR