[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) 95%

Supplier: Ambeed
Warning

Synonyms: Cobalt tetramethoxyphenylporphyrin, [5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinato]cobalt(II), Cobalt(II) meso-tetra(4-methoxyphenyl)porphine, 5,10,15,20-Tetrakis (4-methoxyphenyl)-21H,23H-porphine cobalt (II), 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)

A236788-10G A236788-1G A236788-5G A236788-250MG
76771-288EA 287 USD
76771-288 76771-290 76771-292 77600-904
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) 95%
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II)

Precaución: for Research Use Only

Formula: C₄₈H₃₆CoN₄O₄
MW: 791.77 g/mol
MDL Number: MFCD00010724
CAS Number: 28903-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR