6-Chloro-3-nitroimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A235543-1G A235543-250MG A235543-5G
76771-264EA 15 USD
76771-264 76771-266 76771-268
6-Chloro-3-nitroimidazo[1,2-b]pyridazine 97%
6-Chloro-3-nitroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₃ClN₄O₂
MDL Number: MFCD08276991
CAS Number: 18087-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR