2-Bromo-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A235507-100G A235507-10G A235507-1G A235507-500G A235507-5G
76771-256EA 27 USD
76771-256 76771-258 76771-260 76771-262 77593-514
2-Bromo-4-fluorobenzaldehyde 98%
2-Bromo-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO
MDL Number: MFCD00672923
CAS Number: 59142-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR