Methyl-3,5-dihydroxyphenylacetate 97%

Supplier: Ambeed
Warning

A234990-250MG A234990-5G A234990-1G
76771-248EA 15 USD
76771-248 77550-514 77626-244
Methyl-3,5-dihydroxyphenylacetate 97%
Methyl-3,5-dihydroxyphenylacetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00091620
CAS Number: 4724-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR