6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A234191-100MG A234191-250MG A234191-5G A234191-1G
76771-210EA 16 USD
76771-210 76771-212 76771-214 77600-880
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 97%
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O
MDL Number: MFCD11044635
CAS Number: 171178-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR