5-Phenyl-1,3,4-oxadiazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-phenyl-1,3,4-oxadiazole

A233906-10G A233906-1G A233906-250MG A233906-25G A233906-5G
76771-200EA 80 USD
76771-200 76771-202 76771-204 76771-206 76771-208
5-Phenyl-1,3,4-oxadiazol-2-amine 98%
5-Phenyl-1,3,4-oxadiazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD00126383
CAS Number: 1612-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR