5-Fluoro-2-iodotoluene 97%

Supplier: Ambeed
Warning

A231057-10G A231057-25G
76771-148EA 7.43 USD
76771-148 76771-150
5-Fluoro-2-iodotoluene 97%
5-Fluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆FI
MDL Number: MFCD00039397
CAS Number: 66256-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR