6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol 97%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dihydro-5H-cyclopenta[b]pyridin-7-ol

A229880-1G A229880-250MG
76771-114EA 82.76 USD
76771-114 76771-116
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol 97%
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.163 g/mol
MDL Number: MFCD07778342
CAS Number: 41598-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR