2,6-Dibromo-4-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

A229766-10G A229766-25G A229766-5G A229766-100G A229766-500G
76771-104EA 6 USD
76771-104 76771-106 76771-108 77877-757 77877-758
2,6-Dibromo-4-(trifluoromethoxy)aniline 98%
2,6-Dibromo-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₃NO
MDL Number: MFCD00153113
CAS Number: 88149-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR