6-Hydroxy-2-naphthaldehyde 98%

Supplier: Ambeed
Warning

A229246-10G A229246-1G A229246-25G A229246-5G
76771-090EA 49.87 USD
76771-090 76771-092 76771-094 76771-096
6-Hydroxy-2-naphthaldehyde 98%
6-Hydroxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.1800 g/mol
MDL Number: MFCD03427537
CAS Number: 78119-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR